Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143905.2 | NP_001137377.1 | 3 | 625 | ||
NM_001194995.1 | NP_001181924.1 | 3 | 918 | |||
NM_152269.4 | NP_689482.1 | 3 | 709 | |||
XM_005253630.4 | XP_005253687.1 | 4 | 729 | |||
XM_011538980.2 | XP_011537282.1 | 3 | 1487 | |||
GenBank mRNA | AF061733.1 | - | 3 | 551 | ||
AK095982.1 | - | 3 | 1089 | |||
BC018145.1 | - | 3 | 510 | 3846678 | ||
BC062329.1 | - | 3 | 552 | 5484911 |