Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317899.1 | NP_001304828.1 | 10 | 1411 | ||
NM_152272.4 | NP_689485.1 | 11 | 2125 | |||
XM_006716415.2 | XP_006716478.1 | 9 | 1520 | |||
XM_017013961.1 | XP_016869450.1 | 9 | 1795 | |||
XM_017013962.1 | XP_016869451.1 | 11 | 2476 | |||
XM_017013963.1 | XP_016869452.1 | 9 | 1602 | |||
XM_017013964.1 | XP_016869453.1 | 11 | 3274 | |||
XM_017013965.1 | XP_016869454.1 | 9 | 1327 | |||
GenBank mRNA | AK021776.1 | - | 10 | 1554 | ||
AK092500.1 | - | 10 | 1361 | |||
AK097202.1 | - | 11 | 2122 | |||
AL832313.1 | - | 7 | 1506 | |||
AL833843.1 | - | 10 | 1525 | |||
BC004344.1 | - | 10 | 1353 | |||
BC019110.1 | - | 10 | 1527 | 5093453 | ||
BC042050.1 | - | 11 | 2106 | 5551762 |