Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286139.1 | NP_001273068.1 | 9 | 1191 | ||
NM_152313.3 | NP_689526.2 | 9 | 1154 | |||
XM_017017174.1 | XP_016872663.1 | 9 | 1158 | |||
XM_017017175.1 | XP_016872664.1 | 8 | 1071 | |||
XM_017017176.1 | XP_016872665.1 | 8 | 1023 | |||
XM_017017177.1 | XP_016872666.1 | 8 | 927 | |||
GenBank mRNA | AJ295983.1 | - | 4 | 528 | ||
AK096251.1 | - | 9 | 1190 | |||
AY162216.1 | - | 9 | 1108 | |||
BC047374.1 | - | 9 | 1121 | 5313230 |