Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195736.1 | NP_001182665.1 | 5, 6 | 658 | ||
NM_152371.3 | NP_689584.2 | 5, 6 | 604 | |||
NR_036637.1 | 5, 6 | 537 | ||||
XM_006710354.3 | XP_006710417.1 | 5, 6 | 592 | |||
XM_011540664.1 | XP_011538966.1 | 5, 6 | 648 | |||
GenBank mRNA | AF425266.1 | - | 5, 6 | 548 | ||
AK057027.1 | - | 5, 6 | 589 | |||
AK308034.1 | - | 5, 6 | 514 | |||
BC022547.1 | - | 5, 6 | 544 | 4812314 | ||
BQ709693.1 | - | 440 | ||||
DA495138.1 | - | 5, 6 | 537 |