Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152434.2 | NP_689647.2 | 5 | 571 | ||
XM_011542620.2 | XP_011540922.1 | 5 | 569 | |||
XM_017017239.1 | XP_016872728.1 | 5 | 542 | |||
GenBank mRNA | AK056905.1 | - | 3 | 223 | ||
AK126881.1 | - | 5 | 564 | |||
AK293001.1 | - | 5 | 506 | |||
BC070142.1 | - | 5 | 550 | |||
BC107418.1 | - | 5 | 554 | |||
BC110439.1 | - | 2 | 184 | 40001346 | ||
BC110440.1 | - | 2 | 184 | 40001348 | ||
BC110441.1 | - | 2 | 184 | 40001349 | ||
BC110442.1 | - | 2 | 184 | 40001350 | ||
BC118669.1 | - | 2 | 187 | 40012545 | ||
BX647832.1 | - | 5 | 545 | |||
BX648704.1 | - | 5 | 550 |