Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270396.1 | NP_001257325.1 | 1 | 199 | ||
NM_152436.2 | NP_689649.1 | 1 | 199 | |||
NR_072995.1 | 1 | 199 | ||||
XM_011537949.2 | XP_011536251.1 | 1 | 180 | |||
GenBank mRNA | AK093175.1 | - | 1 | 189 | ||
BC016749.1 | - | 1 | 206 | |||
BC029557.1 | - | 1 | 160 | 5262538 | ||
BX640916.1 | - | 1 | 168 |