Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 15 | 1547 | ||
XM_005273409.1 | XP_005273466.1 | 16 | 1658 | |||
XM_005273410.1 | XP_005273467.1 | 16 | 1634 | |||
XM_005273411.1 | XP_005273468.1 | 14 | 1466 | |||
XM_017013088.1 | XP_016868577.1 | 13 | 1355 | |||
XM_017013089.1 | XP_016868578.1 | 14 | 1420 | |||
GenBank mRNA | AK057293.1 | - | 6 | 937 | ||
AK074839.1 | - | 2 | 117 | |||
AK304441.1 | - | 15 | 1499 | |||
AK308922.1 | - | 1 | 815 | |||
CR749838.1 | - | 11 | 971 |