Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 18 | 2235 | ||
XM_005273409.1 | XP_005273466.1 | 19 | 2346 | |||
XM_005273410.1 | XP_005273467.1 | 19 | 2322 | |||
XM_005273411.1 | XP_005273468.1 | 17 | 2154 | |||
XM_017013088.1 | XP_016868577.1 | 16 | 2043 | |||
XM_017013089.1 | XP_016868578.1 | 17 | 2108 | |||
GenBank mRNA | AK057293.1 | - | 9 | 1625 | ||
AK074839.1 | - | 5 | 805 | |||
AK304441.1 | - | 18 | 2187 | |||
AK308922.1 | - | 4 | 1503 | |||
CR749838.1 | - | 14 | 1659 |