Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 11 | 1318 | ||
NM_001142557.1 | NP_001136029.1 | 8 | 1091 | |||
NM_012484.2 | NP_036616.2 | 11 | 1315 | |||
NM_012485.2 | NP_036617.2 | 10 | 1270 | |||
GenBank mRNA | AF032862.1 | - | 11 | 1169 | ||
AK290577.1 | - | 10 | 1111 | |||
AK302975.1 | - | 12 | 1125 | |||
AK303616.1 | - | 8 | 969 | |||
AK313603.1 | - | 11 | 1236 | |||
AY623426.1 | - | 11 | 1141 | |||
BC057235.1 | - | 10 | 1240 | |||
BC070281.1 | - | 10 | 1146 | |||
BC108904.1 | - | 11 | 1170 | 40007903 | ||
U29343.1 | - | 11 | 1196 |