Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVV-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152473.2 | NP_689686.2 | 1 | 1259 | ||
GenBank mRNA | AK056776.1 | - | 2 | 984 | ||
BC104018.1 | - | 1 | 989 | |||
BC104019.1 | - | 1 | 989 | |||
BC104020.1 | - | 1 | 989 | |||
BC113852.1 | - | 1 | 987 | |||
BC114460.1 | - | 1 | 987 |
Gene Symbol: ERVV-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191055.1 | NP_001177984.1 | 2 | 1865 |