Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152548.2 | NP_689761.2 | 4 | 388 | ||
XM_005271905.2 | XP_005271962.2 | 4 | 366 | |||
XM_005271906.4 | XP_005271963.2 | 5 | 411 | |||
XM_005271907.3 | XP_005271964.1 | 4 | 373 | |||
XM_011543207.1 | XP_011541509.1 | 4 | 383 | |||
XM_011543208.1 | XP_011541510.1 | 4 | 383 | |||
XM_011543209.1 | XP_011541511.1 | 2 | 69 | |||
XM_011543210.2 | XP_011541512.1 | 2 | 221 | |||
XM_017009118.1 | XP_016864607.1 | 5 | 408 | |||
GenBank mRNA | AK058062.1 | - | 4 | 371 | ||
AK310245.1 | - | 3 | 190 | |||
BC034772.1 | - | 4 | 388 | 5169904 | ||
BM560335.1 | - | 3 | 193 |