Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152589.1 | NP_689802.1 | 9 | 884 | ||
XM_017018886.1 | XP_016874375.1 | 8 | 824 | |||
XM_017018887.1 | XP_016874376.1 | 9 | 1127 | |||
GenBank mRNA | AK093140.1 | - | 9 | 884 | ||
BC038413.2 | - | 8 | 825 | 5170365 |