Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113523.2 | NP_001106995.1 | 4, 5 | 845 | ||
NM_001308321.1 | NP_001295250.1 | 1 | 172 | |||
NM_152615.2 | NP_689828.1 | 1 | 172 | |||
XM_005247159.3 | XP_005247216.1 | 4, 5 | 2517 | |||
XM_005247160.3 | XP_005247217.1 | 3, 4 | 2289 | |||
XM_011512475.2 | XP_011510777.1 | 3, 4 | 2401 | |||
XM_011512480.2 | XP_011510782.1 | 4, 5 | 2533 | |||
XM_017005791.1 | XP_016861280.1 | 3, 4 | 2289 | |||
GenBank mRNA | AK097916.1 | - | 1 | 109 | ||
BC101701.1 | - | 1 | 172 | 8069207 | ||
BC101703.1 | - | 1 | 172 | 8069209 | ||
BC143573.1 | - | 1 | 172 | 9052084 | ||
DQ063586.1 | - | 4, 5 | 764 |