Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317906.1 | NP_001304835.1 | 14 | 2839 | ||
NM_001317907.1 | NP_001304836.1 | 15 | 2775 | |||
NM_152562.3 | NP_689775.2 | 15 | 2756 | |||
XM_006716294.2 | XP_006716357.1 | 15 | 2771 | |||
XM_011544415.1 | XP_011542717.1 | 16 | 2843 | |||
XM_011544416.2 | XP_011542718.1 | 16 | 2537 | |||
XM_011544417.2 | XP_011542719.1 | 16 | 2594 | |||
XM_011544418.1 | XP_011542720.1 | 16 | 2807 | |||
GenBank mRNA | AK092164.1 | - | 10 | 1372 | ||
AK098670.1 | - | 2 | 533 | |||
AK292505.1 | - | 14 | 2571 | |||
AL833396.1 | - | 12 | 1861 | |||
AL833627.1 | - | 14 | 2533 | |||
BC036214.1 | - | 12 | 1884 | 5267276 | ||
BC063651.1 | - | 2 | 464 | |||
BC085609.1 | - | 2 | 519 | |||
BG354575.1 | - | 2 | 540 | |||
BX649008.1 | - | 11 | 1747 |