Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF567 SIRNA Assays ›
Gene Symbol:
ZNF567
Gene Name:
zinc finger protein 567
Gene Aliases:
-
Chromosome Location:
Chr.19: 36666961 - 36723548 on Build GRCh38
UniGene ID:
Hs.412517
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39376
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF567
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300979.1
NP_001287908.1
6
1037
NM_001322911.1
NP_001309840.1
5
910
NM_001322912.1
NP_001309841.1
5
962
NM_001322913.1
NP_001309842.1
6
1041
NM_001322914.1
NP_001309843.1
6
1213
NM_001322915.1
NP_001309844.1
7
1109
NM_001322916.1
NP_001309845.1
6
985
NM_001322917.1
NP_001309846.1
6
989
NM_001322918.1
NP_001309847.1
7
1113
NM_001322919.1
NP_001309848.1
6
1209
NM_001322920.1
NP_001309849.1
7
1161
NM_152603.4
NP_689816.2
5
914
XM_011526584.2
XP_011524886.1
4
841
XM_011526585.2
XP_011524887.1
4
841
XM_017026416.1
XP_016881905.1
5
1173
XM_017026417.1
XP_016881906.1
8
2256
XM_017026418.1
XP_016881907.1
6
1116
XM_017026419.1
XP_016881908.1
5
1172
XM_017026420.1
XP_016881909.1
6
2025
XM_017026421.1
XP_016881910.1
5
1136
GenBank mRNA
AK024149.1
-
1
41
AK093034.1
-
5
846
AK126691.1
-
3
1936
AK298556.1
-
5
904
AK303326.1
-
6
1021
AK313647.1
-
5
910
BC033849.2
-
4
888
4472579
BX640706.1
-
6
1026
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0