Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152608.3 | NP_689821.3 | 6 | 1084 | ||
GenBank mRNA | AK092701.1 | - | 6 | 910 | ||
AK127780.1 | - | 5 | 1014 | |||
AK291008.1 | - | 6 | 1047 | |||
BC071563.1 | - | 6 | 1062 | 5261192 | ||
BX648319.1 | - | 6 | 1042 |