Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113523.2 | NP_001106995.1 | 6 | 1054 | ||
NM_001308320.1 | NP_001295249.1 | 4 | 676 | |||
NM_001308321.1 | NP_001295250.1 | 2 | 381 | |||
NM_152615.2 | NP_689828.1 | 2 | 381 | |||
XM_005247159.3 | XP_005247216.1 | 6 | 2726 | |||
XM_005247160.3 | XP_005247217.1 | 5 | 2498 | |||
XM_011512475.2 | XP_011510777.1 | 5 | 2610 | |||
XM_011512476.2 | XP_011510778.1 | 5 | 2579 | |||
XM_011512477.2 | XP_011510779.1 | 4 | 2348 | |||
XM_011512478.2 | XP_011510780.1 | 6 | 837 | |||
XM_011512479.2 | XP_011510781.1 | 4 | 312 | |||
XM_011512480.2 | XP_011510782.1 | 6 | 2742 | |||
XM_017005791.1 | XP_016861280.1 | 5 | 2498 | |||
XM_017005792.1 | XP_016861281.1 | 3 | 1479 | |||
GenBank mRNA | AK097515.1 | - | 8 | 999 | ||
AK097916.1 | - | 2 | 318 | |||
BC101701.1 | - | 2 | 381 | 8069207 | ||
BC101703.1 | - | 2 | 381 | 8069209 | ||
BC143573.1 | - | 2 | 381 | 9052084 | ||
BX647656.1 | - | 4 | 652 | |||
DQ063586.1 | - | 6 | 973 |