Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 2 | 412 | ||
NM_001291833.1 | NP_001278762.1 | 2 | 406 | |||
NM_020790.1 | NP_065841.1 | 2 | 409 | |||
NM_152673.3 | NP_689886.3 | 2 | 412 | |||
GenBank mRNA | AB037780.1 | - | 1 | 2050 | ||
AB098731.1 | - | 1 | 663 | |||
AK027314.1 | - | 2 | 316 | |||
AK075138.1 | - | 2 | 349 | |||
AK298316.1 | - | 2 | 534 | |||
AK311504.1 | - | 2 | 410 | |||
AY358449.1 | - | 2 | 345 | |||
BC029267.1 | - | 2 | 320 | 3851952 | ||
BC044243.1 | - | 2 | 299 | 5754328 | ||
BM721408.1 | - | 2 | 240 | |||
DC348309.1 | - | 2 | 412 |