Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152740.3 | NP_689953.1 | 2, 3 | 465 | ||
GenBank mRNA | AB050000.1 | - | 2, 3 | 339 | ||
AF529362.1 | - | 2, 3 | 252 | |||
AK025558.1 | - | 1, 2 | 77 | |||
AK316605.1 | - | 2, 3 | 270 | |||
BC020167.1 | - | 2, 3 | 326 | |||
BC032324.1 | - | 2, 3 | 350 | 3890576 | ||
BP343005.1 | - | 2, 3 | 465 | |||
BQ051350.1 | - | 447 |