Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSGA10IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152762.2 | NP_689975.2 | 2 | 252 | ||
XM_011544887.2 | XP_011543189.1 | 2 | 451 | |||
XM_011544888.2 | XP_011543190.1 | 2 | 249 | |||
GenBank mRNA | AK057442.1 | - | 2 | 252 | ||
BC104022.1 | - | 2 | 252 | 40025091 |