Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307922.1 | NP_001294851.1 | 6 | 761 | ||
NM_152682.3 | NP_689895.2 | 6 | 765 | |||
GenBank mRNA | AK021702.1 | - | 6 | 690 | ||
AK293274.1 | - | 4 | 506 | |||
AK315515.1 | - | 6 | 699 | |||
AL540937.3 | - | 6 | 702 | |||
AL559547.3 | - | 6 | 689 | |||
BC017472.1 | - | 6 | 700 | 3909581 | ||
BC067752.1 | - | 6 | 724 | 5270461 | ||
BC107432.1 | - | 6 | 546 | 6104330 |