Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRIMPOL SIRNA Assays ›
Gene Symbol:
PRIMPOL
Gene Name:
primase and DNA directed polymerase
Gene Aliases:
CCDC111, MYP22
Chromosome Location:
Chr.4: 184649613 - 184694959 on Build GRCh38
UniGene ID:
Hs.481307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39536
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRIMPOL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300767.1
NP_001287696.1
6
913
NM_001300768.1
NP_001287697.1
7
1152
NM_152683.3
NP_689896.1
7
1152
XM_011531719.1
XP_011530021.1
7
1029
XM_011531720.1
XP_011530022.1
7
1067
XM_011531721.2
XP_011530023.1
7
1029
XM_011531723.1
XP_011530025.1
6
854
XM_011531724.2
XP_011530026.1
5
811
XM_011531725.1
XP_011530027.1
5
725
XM_011531726.2
XP_011530028.1
4
572
XM_011531729.1
XP_011530031.1
6
858
XM_011531730.2
XP_011530032.1
6
873
XM_017007864.1
XP_016863353.1
7
1067
XM_017007865.1
XP_016863354.1
7
1067
XM_017007866.1
XP_016863355.1
7
1067
XM_017007867.1
XP_016863356.1
7
1029
XM_017007868.1
XP_016863357.1
7
1029
XM_017007869.1
XP_016863358.1
7
1029
XM_017007870.1
XP_016863359.1
7
1029
XM_017007871.1
XP_016863360.1
5
810
XM_017007872.1
XP_016863361.1
5
720
XM_017007873.1
XP_016863362.1
6
899
XM_017007874.1
XP_016863363.1
5
718
XM_017007875.1
XP_016863364.1
6
858
GenBank mRNA
AK057729.1
-
7
935
AK307317.1
-
6
858
BC047467.1
-
6
859
BC064600.1
-
7
993
5786393
BX647575.1
-
7
1040
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial DNA replication
DNA replication, synthesis of RNA primer
response to UV
translesion synthesis
replication fork processing
chromatin binding
DNA-directed DNA polymerase activity
DNA primase activity
protein binding
manganese ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0