Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152727.5 | NP_689940.3 | 3 | 552 | ||
GenBank mRNA | AF492484.1 | - | 3 | 503 | ||
AK292269.1 | - | 2 | 461 | |||
AK315394.1 | - | 3 | 455 | |||
BC053658.1 | - | 4 | 626 | 5750192 | ||
BG830959.1 | - | 3 | 498 | |||
BG831872.1 | - | 3 | 500 | |||
CR749617.1 | - | 2 | 213 |