Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098519.1 | NP_001091989.1 | 6 | 1087 | ||
NM_152759.4 | NP_689972.3 | 6 | 976 | |||
XM_017019124.1 | XP_016874613.1 | 6 | 1095 | |||
GenBank mRNA | AK124107.1 | - | 8 | 1356 | ||
BC029245.2 | - | 5 | 911 | 5169080 |