Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307922.1 | NP_001294851.1 | 8 | 973 | ||
NM_152682.3 | NP_689895.2 | 8 | 977 | |||
GenBank mRNA | AK021702.1 | - | 8 | 902 | ||
AK293274.1 | - | 6 | 718 | |||
AL540937.3 | - | 7 | 913 | |||
BC017472.1 | - | 8 | 912 | 3909581 | ||
BC067752.1 | - | 8 | 936 | 5270461 | ||
BC107432.1 | - | 8 | 758 | 6104330 |