Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 8 | 1717 | ||
XM_011525875.2 | XP_011524177.1 | 8 | 1753 | |||
GenBank mRNA | AK057795.1 | - | 7 | 1426 | ||
AY599248.1 | - | 8 | 1715 | |||
BC008583.1 | - | 6 | 1299 | |||
BC019045.2 | - | 6 | 1342 | |||
BC096744.1 | - | 8 | 1710 | 5260167 |