Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098519.1 | NP_001091989.1 | 10, 11 | 1841 | ||
NM_152759.4 | NP_689972.3 | 10, 11 | 1730 | |||
XM_017019124.1 | XP_016874613.1 | 10, 11 | 1849 | |||
GenBank mRNA | AK124107.1 | - | 12, 13 | 2110 | ||
BC029245.2 | - | 9, 10 | 1665 | 5169080 |