Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152769.2 | NP_689982.3 | 9 | 1419 | ||
XM_017026556.1 | XP_016882045.1 | 9 | 1393 | |||
XM_017026557.1 | XP_016882046.1 | 8 | 1840 | |||
XM_017026558.1 | XP_016882047.1 | 8 | 983 | |||
XM_017026559.1 | XP_016882048.1 | 9 | 1538 | |||
GenBank mRNA | BC028156.1 | - | 9 | 1270 | 5241080 |