Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152889.2 | NP_690849.1 | 3 | 1359 | ||
GenBank mRNA | AY120869.1 | - | 3 | 1314 | ||
BC039390.1 | - | 1 | 503 | |||
BC045257.1 | - | 1 | 523 | |||
BC047236.1 | - | 1 | 523 | |||
BC103895.1 | - | 3 | 1314 | 40005100 | ||
BC103896.1 | - | 4 | 1133 | 40005101 | ||
BC103897.1 | - | 4 | 1133 | 40005103 |