Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADGRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294335.1 | NP_001281264.1 | 4 | 543 | ||
NM_001294336.1 | NP_001281265.1 | 4 | 543 | |||
XM_011541848.2 | XP_011540150.1 | 3 | 497 | |||
XM_011541849.2 | XP_011540151.1 | 2 | 332 | |||
XM_017001899.1 | XP_016857388.1 | 3 | 627 | |||
XM_017001900.1 | XP_016857389.1 | 2 | 599 | |||
XM_017001901.1 | XP_016857390.1 | 2 | 599 | |||
XM_017001902.1 | XP_016857391.1 | 2 | 600 | |||
XM_017001903.1 | XP_016857392.1 | 2 | 600 | |||
XM_017001904.1 | XP_016857393.1 | 2 | 596 | |||
XM_017001905.1 | XP_016857394.1 | 2 | 598 | |||
XM_017001906.1 | XP_016857395.1 | 2 | 597 | |||
XM_017001907.1 | XP_016857396.1 | 2 | 596 | |||
XM_017001908.1 | XP_016857397.1 | 2 | 597 | |||
XM_017001909.1 | XP_016857398.1 | 2 | 598 | |||
XM_017001910.1 | XP_016857399.1 | 2 | 596 | |||
XM_017001911.1 | XP_016857400.1 | 2 | 597 | |||
XM_017001912.1 | XP_016857401.1 | 2 | 595 | |||
GenBank mRNA | AB005298.1 | - | 4 | 515 | ||
BC136533.1 | - | 2 | 211 | 9020523 | ||
DA314854.1 | - | 2 | 316 | |||
DA494051.1 | - | 2 | 332 |
Set Membership: |
G-protein coupled receptor |