Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152889.2 | NP_690849.1 | 3 | 1373 | ||
GenBank mRNA | AY120869.1 | - | 3 | 1328 | ||
BC039390.1 | - | 1 | 517 | |||
BC045257.1 | - | 1 | 537 | |||
BC047236.1 | - | 1 | 537 |