Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024008.1 | 1 | 1416 | |||
GenBank mRNA | AK054839.1 | - | 1 | 1416 | ||
BC126477.1 | - | 1 | 962 | |||
BC126479.1 | - | 1 | 962 |