Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267576.1 | NP_001254505.1 | 15 | 2827 | ||
NM_153210.4 | NP_694942.3 | 15 | 2842 | |||
XM_011523639.2 | XP_011521941.1 | 14 | 2730 | |||
XM_011523640.1 | XP_011521942.1 | 14 | 2562 | |||
XM_011523641.1 | XP_011521943.1 | 15 | 2287 | |||
XM_011523642.2 | XP_011521944.1 | 15 | 2887 | |||
XM_017024159.1 | XP_016879648.1 | 14 | 2783 | |||
XM_017024160.1 | XP_016879649.1 | 13 | 2769 | |||
XM_017024161.1 | XP_016879650.1 | 12 | 2855 | |||
XM_017024162.1 | XP_016879651.1 | 15 | 2872 | |||
GenBank mRNA | AJ583817.2 | - | 15 | 2749 | ||
AK055188.1 | - | 15 | 2321 | |||
AK090821.1 | - | 4 | 1321 | |||
BC039653.1 | - | 4 | 804 | |||
BC136368.1 | - | 15 | 2827 | 9020355 | ||
BC144041.1 | - | 15 | 2842 | 9052561 | ||
BC171759.1 | - | 15 | 2827 | 9054412 | ||
BX648526.1 | - | 13 | 2105 |