Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144972.1 | NP_001138444.1 | 3 | 439 | ||
NM_153337.2 | NP_699168.1 | 3 | 439 | |||
NM_182854.3 | NP_878274.1 | 3 | 439 | |||
GenBank mRNA | AK055837.1 | - | 3 | 404 | ||
AK292650.1 | - | 3 | 398 | |||
AK292705.1 | - | 3 | 404 | |||
AY302441.1 | - | 3 | 341 | |||
BC027944.1 | - | 3 | 388 | |||
BC063423.1 | - | 3 | 380 | |||
BC119626.1 | - | 3 | 292 |