Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 4 | 383 | ||
XM_017023023.1 | XP_016878512.1 | 4 | 383 | |||
GenBank mRNA | AB067511.1 | - | 3 | 771 | ||
AK091062.1 | - | 4 | 383 | |||
BC121185.1 | - | 4 | 393 | |||
BC121186.1 | - | 4 | 393 |