Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 574 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 685 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 598 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 896 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 577 | |||
NM_153345.3 | NP_699176.1 | 3 | 476 | |||
NR_040003.2 | 2 | 634 | ||||
NR_104250.1 | 3 | 315 | ||||
NR_104251.1 | 2 | 214 | ||||
NR_104252.1 | 3 | 312 | ||||
NR_104253.1 | 2 | 238 | ||||
NR_104254.1 | 3 | 336 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 421 | ||
AK172816.1 | - | 5 | 627 | |||
AK315170.1 | - | 4 | 522 | |||
AY358781.1 | - | 3 | 491 | |||
BC035517.1 | - | 3 | 876 | 5207904 | ||
BC035888.1 | - | 2 | 237 | |||
BE562464.1 | - | 2 | 221 | |||
BF036500.1 | - | 1 | 134 | |||
BI761527.1 | - | 2 | 164 | |||
CB142336.1 | - | 3 | 263 | |||
CB156055.1 | - | 2 | 154 | |||
DB222115.1 | - | 3 | 255 | |||
DT220400.1 | - | 2 | 166 | |||
R69052.1 | - | 3 | 261 |