Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153367.3 | NP_699198.2 | 4 | 1185 | ||
XM_011539452.2 | XP_011537754.1 | 4 | 1136 | |||
GenBank mRNA | AK075279.1 | - | 4 | 1037 | ||
AK293361.1 | - | 4 | 824 | |||
AK303734.1 | - | 4 | 816 | |||
AK308876.1 | - | 5 | 1116 | |||
BC028617.1 | - | 4 | 1187 | 4821347 | ||
BC039249.1 | - | 4 | 1179 | |||
BC053564.1 | - | 4 | 1084 | 6193781 | ||
BX648607.1 | - | 1 | 71 |