Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000576.2 | NP_000567.1 | 5 | 539 | ||
XM_017003988.1 | XP_016859477.1 | 4 | 540 | |||
GenBank mRNA | AB451494.1 | - | 1 | 104 | ||
AF043335.1 | - | 1 | 104 | |||
AK313983.1 | - | 4 | 1259 | |||
BC008678.1 | - | 5 | 539 | 3875593 | ||
BT007213.1 | - | 4 | 452 | |||
CR407679.1 | - | 4 | 452 | |||
K02770.1 | - | 5 | 538 | |||
M15330.1 | - | 5 | 538 | |||
M54933.1 | - | 5 | 539 | |||
X02532.1 | - | 5 | 529 | |||
X56087.1 | - | 5 | 509 |