Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153267.4 | NP_694999.3 | 14 | 2847 | ||
NR_125850.1 | 14 | 2843 | ||||
GenBank mRNA | BC015417.2 | - | 6 | 911 | ||
BC016383.1 | - | 14 | 2565 | 4696722 | ||
BC063634.1 | - | 14 | 2824 | 4399866 | ||
BX538309.1 | - | 3 | 3838 |