Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 4 | 1700 | ||
NM_001242774.2 | NP_001229703.1 | 5 | 1811 | |||
NM_001242775.2 | NP_001229704.1 | 4 | 1724 | |||
NM_001282876.1 | NP_001269805.1 | 3 | 2022 | |||
NM_001282877.1 | NP_001269806.1 | 4 | 1703 | |||
NM_153345.3 | NP_699176.1 | 3 | 1602 | |||
NR_040003.2 | 2 | 1760 | ||||
NR_104250.1 | 3 | 1441 | ||||
NR_104251.1 | 2 | 1340 | ||||
NR_104252.1 | 3 | 1438 | ||||
NR_104253.1 | 2 | 1364 | ||||
NR_104254.1 | 3 | 1462 | ||||
GenBank mRNA | AK075067.1 | - | 3 | 1547 | ||
AK172816.1 | - | 5 | 1754 | |||
BC035517.1 | - | 3 | 2002 | 5207904 | ||
BC035888.1 | - | 2 | 1363 |