Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103534.1 | 5 | 907 | |||
GenBank mRNA | BC030279.1 | - | 2 | 538 | 5141008 |