Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244889.1 | NP_001231818.1 | 4, 5 | 1011 | ||
NM_170744.4 | NP_734465.2 | 4, 5 | 1011 | |||
XM_011539453.1 | XP_011537755.1 | 5, 6 | 560 | |||
XM_017015834.1 | XP_016871323.1 | 5, 6 | 570 | |||
XM_017015835.1 | XP_016871324.1 | 4, 5 | 1363 | |||
GenBank mRNA | AB096256.1 | - | 4, 5 | 966 | ||
AK128132.1 | - | 4, 5 | 1011 | |||
AY126437.1 | - | 4, 5 | 619 | |||
AY358351.1 | - | 4, 5 | 957 |