Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153610.4 | NP_705838.3 | 2 | 8458 | ||
XM_017009212.1 | XP_016864701.1 | 2 | 8326 | |||
GenBank mRNA | AL831966.1 | - | 1 | 591 | ||
AL832021.1 | - | 1 | 832 | |||
AL832368.1 | - | 1 | 66 | |||
AL834252.2 | - | 1 | 175 | |||
BC046215.1 | - | 1 | 1095 | |||
BC062664.1 | - | 1 | 612 | |||
BX647452.1 | - | 1 | 4604 | |||
BX647628.1 | - | 1 | 162 | |||
BX647646.1 | - | 1 | 162 |