Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170743.3 | NP_734464.1 | 3 | 269 | ||
NM_173064.2 | NP_775087.1 | 3 | 269 | |||
NM_173065.2 | NP_775088.1 | 3 | 269 | |||
XM_006710394.3 | XP_006710457.2 | 2 | 333 | |||
XM_017000479.1 | XP_016855968.1 | 2 | 298 | |||
GenBank mRNA | AF439325.1 | - | 3 | 242 | ||
AJ534330.1 | - | 3 | 255 | |||
AJ534331.1 | - | 3 | 255 | |||
AK160364.1 | - | 3 | 245 | |||
AY129151.1 | - | 3 | 242 | |||
AY129152.1 | - | 3 | 242 | |||
AY129153.1 | - | 3 | 242 | |||
BC101407.1 | - | 2 | 131 | |||
BC101408.1 | - | 2 | 131 | |||
BC101975.1 | - | 3 | 255 | |||
BC139722.1 | - | 3 | 255 | |||
BC140872.1 | - | 3 | 318 | 9021740 | ||
CN289861.1 | - | 3 | 269 | |||
JN591376.1 | - | 3 | 242 |