Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40506
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
860
NM_001143806.1
NP_001137278.1
2
1075
NM_001143807.1
NP_001137279.1
2
961
NM_001143808.1
NP_001137280.1
2
797
NM_001143809.1
NP_001137281.1
2
807
NM_001143810.1
NP_001137282.1
3
1090
NM_001143811.1
NP_001137283.1
4
1207
NM_001143812.1
NP_001137284.1
2
949
NM_001143813.1
NP_001137285.1
2
1031
NM_001143814.1
NP_001137286.1
3
1156
NM_001143816.1
NP_001137288.1
2
1554
NM_001709.4
NP_001700.2
2
1049
NM_170731.4
NP_733927.1
2
1371
NM_170732.4
NP_733928.1
2
1158
NM_170733.3
NP_733929.1
2
1061
NM_170734.3
NP_733930.1
2
1009
NM_170735.5
NP_733931.1
1
1788
XM_011520280.2
XP_011518582.1
2
1955
GenBank mRNA
AF400438.1
-
1
721
AK289763.1
-
2
857
AK289853.1
-
2
758
AY054392.2
-
2
1062
AY054393.2
-
2
804
AY054394.2
-
2
1019
AY054395.2
-
2
1102
AY054396.2
-
2
860
AY054397.2
-
2
943
AY054398.2
-
2
990
AY054399.2
-
3
875
AY054400.2
-
2
976
AY054406.3
-
3
700
AY656701.1
-
1
703
BC029795.1
-
2
1034
5193877
EF674517.1
-
2
860
EF674518.1
-
2
1075
EF674519.1
-
2
1158
EF674520.1
-
2
961
EF674521.1
-
2
1061
EF689009.1
-
1
1802
EF689010.1
-
2
1568
EF689011.1
-
2
797
EF689012.1
-
2
807
EF689013.1
-
2
949
EF689014.1
-
2
1031
EF689015.1
-
2
1049
EF689016.1
-
3
1156
EF689017.1
-
3
908
EF689018.1
-
2
1009
EF689019.1
-
3
1090
EF689020.1
-
4
1207
EF689021.1
-
2
1371
KP229412.1
-
1
703
KU177943.1
-
1
703
KU177944.1
-
2
586
KU177945.1
-
2
559
M61176.1
-
1
1265
X60201.1
-
2
999
X91251.1
-
2
987
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: AY176065.
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0