Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 34 | 4860 | ||
XM_005246299.4 | XP_005246356.1 | 35 | 4950 | |||
XM_005246300.4 | XP_005246357.1 | 35 | 4893 | |||
XM_006712268.3 | XP_006712331.1 | 35 | 4947 | |||
XM_006712269.3 | XP_006712332.1 | 34 | 4872 | |||
XM_006712270.3 | XP_006712333.2 | 34 | 4806 | |||
XM_006712271.3 | XP_006712334.1 | 34 | 4913 | |||
GenBank mRNA | AB095944.1 | - | 5 | 970 | ||
AK126998.1 | - | 12 | 1589 | |||
AK127553.1 | - | 8 | 866 | |||
AK128490.1 | - | 18 | 2340 | |||
AK303062.1 | - | 9 | 960 | |||
AL834144.1 | - | 6 | 567 | |||
BX649036.1 | - | 10 | 1096 | |||
CR749469.1 | - | 12 | 1362 |