Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 27, 28 | 3261 | ||
XM_011513952.1 | XP_011512254.1 | 27, 28 | 3247 | |||
XM_011513953.1 | XP_011512255.1 | 26, 27 | 3185 | |||
GenBank mRNA | AK097562.1 | - | 3, 4 | 307 | ||
AK126005.1 | - | 12, 13 | 1130 | |||
AK128183.1 | - | 17, 18 | 1727 | |||
AL832763.1 | - | 22, 23 | 2186 | |||
BC044937.1 | - | 15, 16 | 1733 | |||
BC052962.1 | - | 27, 28 | 3222 | 4838071 | ||
CR749346.1 | - | 22, 23 | 2162 |