Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 38 | 5445 | ||
XM_005246299.4 | XP_005246356.1 | 39 | 5535 | |||
XM_005246300.4 | XP_005246357.1 | 39 | 5478 | |||
XM_006712268.3 | XP_006712331.1 | 39 | 5532 | |||
XM_006712269.3 | XP_006712332.1 | 38 | 5457 | |||
XM_006712270.3 | XP_006712333.2 | 38 | 5391 | |||
XM_006712271.3 | XP_006712334.1 | 38 | 5498 | |||
GenBank mRNA | AB095944.1 | - | 9 | 1555 | ||
AK094741.1 | - | 2 | 217 | |||
AK126998.1 | - | 16 | 2174 | |||
AK127553.1 | - | 12 | 1451 | |||
AK128490.1 | - | 22 | 2925 | |||
AK303062.1 | - | 13 | 1545 | |||
AL834144.1 | - | 10 | 1152 | |||
BX649036.1 | - | 14 | 1681 | |||
CR749469.1 | - | 16 | 1947 |