Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172312.1 | NP_758516.1 | 8 | 1693 | ||
GenBank mRNA | BC019247.1 | - | 8 | 1598 | 4825175 | |
S83157.1 | - | 7 | 2429 | |||
U12978.2 | - | 7 | 2429 |