Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308238.1 | NP_001295167.1 | 9 | 1871 | ||
NM_173505.3 | NP_775776.2 | 10 | 1970 | |||
XM_011525830.2 | XP_011524132.1 | 9 | 1890 | |||
XM_011525831.2 | XP_011524133.1 | 9 | 3910 | |||
XM_011525832.2 | XP_011524134.1 | 9 | 4028 | |||
XM_017025561.1 | XP_016881050.1 | 8 | 1794 | |||
GenBank mRNA | AK123197.1 | - | 7 | 3290 | ||
BC030622.1 | - | 10 | 1975 | 4818619 |